Product Details
- SNP ID
-
rs41281644
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:101722939 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTCGGGACCGTTTCCCCAACATC[A/G]TGCAAAGATGGTTCTCGGTACATGG
- Phenotype
-
MIM: 601515
MIM: 604234
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FGF14
PubMed Links
Gene Details
- Gene
- FGF14
- Gene Name
- fibroblast growth factor 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001321931.1 |
983 |
Silent Mutation |
CAC,CAT |
H128H |
NP_001308860.1 |
NM_001321932.1 |
983 |
Silent Mutation |
CAC,CAT |
H149H |
NP_001308861.1 |
NM_001321933.1 |
983 |
Silent Mutation |
CAC,CAT |
H152H |
NP_001308862.1 |
NM_001321934.1 |
983 |
Silent Mutation |
CAC,CAT |
H128H |
NP_001308863.1 |
NM_001321935.1 |
983 |
Silent Mutation |
CAC,CAT |
H128H |
NP_001308864.1 |
NM_001321936.1 |
983 |
Silent Mutation |
CAC,CAT |
H149H |
NP_001308865.1 |
NM_001321937.1 |
983 |
Intron |
|
|
NP_001308866.1 |
NM_001321938.1 |
983 |
Silent Mutation |
CAC,CAT |
H152H |
NP_001308867.1 |
NM_001321939.1 |
983 |
Silent Mutation |
CAC,CAT |
H180H |
NP_001308868.1 |
NM_001321940.1 |
983 |
Silent Mutation |
CAC,CAT |
H152H |
NP_001308869.1 |
NM_001321941.1 |
983 |
Silent Mutation |
CAC,CAT |
H150H |
NP_001308870.1 |
NM_001321942.1 |
983 |
Silent Mutation |
CAC,CAT |
H128H |
NP_001308871.1 |
NM_001321943.1 |
983 |
Silent Mutation |
CAC,CAT |
H128H |
NP_001308872.1 |
NM_001321944.1 |
983 |
Silent Mutation |
CAC,CAT |
H149H |
NP_001308873.1 |
NM_001321945.1 |
983 |
Silent Mutation |
CAC,CAT |
H178H |
NP_001308874.1 |
NM_001321946.1 |
983 |
Silent Mutation |
CAC,CAT |
H128H |
NP_001308875.1 |
NM_001321947.1 |
983 |
Silent Mutation |
CAC,CAT |
H165H |
NP_001308876.1 |
NM_001321948.1 |
983 |
Silent Mutation |
CAC,CAT |
H178H |
NP_001308877.1 |
NM_001321949.1 |
983 |
Silent Mutation |
CAC,CAT |
H128H |
NP_001308878.1 |
NM_004115.3 |
983 |
Silent Mutation |
CAC,CAT |
H212H |
NP_004106.1 |
NM_175929.2 |
983 |
Silent Mutation |
CAC,CAT |
H217H |
NP_787125.1 |
- Gene
- ITGBL1
- Gene Name
- integrin subunit beta like 1
There are no transcripts associated with this gene.
View Full Product Details