Product Details
- SNP ID
-
rs45623838
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:40701142 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTGTGGAGGCTGTTGCCTATGCG[C/T]CAAAGAAGGAGCTAATAAATATTAA
- Phenotype
-
MIM: 179617
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RAD51
PubMed Links
Gene Details
- Gene
- RAD51
- Gene Name
- RAD51 recombinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001164269.1 |
215 |
Missense Mutation |
CCA,TCA |
P56S |
NP_001157741.1 |
NM_001164270.1 |
215 |
Missense Mutation |
CCA,TCA |
P56S |
NP_001157742.1 |
NM_002875.4 |
215 |
Missense Mutation |
CCA,TCA |
P56S |
NP_002866.2 |
NM_133487.3 |
215 |
Missense Mutation |
CCA,TCA |
P56S |
NP_597994.3 |
XM_006720626.3 |
215 |
Missense Mutation |
CCA,TCA |
P56S |
XP_006720689.1 |
XM_011521857.2 |
215 |
Missense Mutation |
CCA,TCA |
P56S |
XP_011520159.1 |
XM_011521858.2 |
215 |
Missense Mutation |
CCA,TCA |
P56S |
XP_011520160.1 |
XM_011521859.2 |
215 |
Missense Mutation |
CCA,TCA |
P56S |
XP_011520161.1 |
XM_011521860.2 |
215 |
Missense Mutation |
CCA,TCA |
P56S |
XP_011520162.1 |
XM_011521861.2 |
215 |
Missense Mutation |
CCA,TCA |
P56S |
XP_011520163.1 |
XM_011521862.2 |
215 |
UTR 5 |
|
|
XP_011520164.1 |
- Gene
- RAD51-AS1
- Gene Name
- RAD51 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details