Product Details

SNP ID
rs45565238
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74749955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTCAAGGATGAGCCAGCGCTAC[A/G]GGGACGTCCTGCAGATCCGCATTGG
Phenotype
MIM: 124060
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CYP1A2 PubMed Links

Gene Details

Gene
CYP1A2
Gene Name
cytochrome P450 family 1 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000761.4 281 Missense Mutation AGG,GGG R73G NP_000752.2

View Full Product Details