Product Details

SNP ID
rs45441395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15950908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAAGTCATCCTCCAGGGCCCCA[C/T]TGGAGGGTTTGGTTTTGTTTGTTTG
Phenotype
MIM: 158343
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCC1 PubMed Links

Gene Details

Gene
ABCC1
Gene Name
ATP binding cassette subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004996.3 Intron NP_004987.2
XM_011522497.1 Intron XP_011520799.1
XM_011522498.2 Intron XP_011520800.1
XM_017023237.1 Intron XP_016878726.1
XM_017023238.1 Intron XP_016878727.1
XM_017023239.1 Intron XP_016878728.1
XM_017023240.1 Intron XP_016878729.1
XM_017023241.1 Intron XP_016878730.1
XM_017023242.1 Intron XP_016878731.1
XM_017023243.1 Intron XP_016878732.1

View Full Product Details