Product Details

SNP ID
rs41359544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58028024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATTTCAAGTCACAGTCCCACCA[C/T]TGAGTTTCTAATCTACTTAGGGATG
Phenotype
MIM: 602261 MIM: 613276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP15 PubMed Links

Gene Details

Gene
MMP15
Gene Name
matrix metallopeptidase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002428.3 Intron NP_002419.1
Gene
USB1
Gene Name
U6 snRNA biogenesis phosphodiesterase 1
There are no transcripts associated with this gene.

View Full Product Details