Product Details

SNP ID
rs45588934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69713556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAAATGAAGACAGGCCTTCTTT[G/T]GTTTTTATTATGGAAATTTTTAACA
Phenotype
MIM: 604708 MIM: 125860
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NFAT5 PubMed Links

Gene Details

Gene
NFAT5
Gene Name
nuclear factor of activated T-cells 5, tonicity-responsive
There are no transcripts associated with this gene.

Gene
NQO1
Gene Name
NAD(P)H quinone dehydrogenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000903.2 Intron NP_000894.1
NM_001025433.1 Intron NP_001020604.1
NM_001025434.1 Intron NP_001020605.1
NM_001286137.1 Intron NP_001273066.1

View Full Product Details