Product Details

SNP ID
rs1804437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5433161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAATCCATTAGGTGGTCATATA[G/A]GGCCTGCAAAGAACAATATTTACTA
Phenotype
MIM: 601269 MIM: 614405
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
C1QBP PubMed Links

Gene Details

Gene
C1QBP
Gene Name
complement C1q binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001212.3 Intron NP_001203.1
Gene
DHX33
Gene Name
DEAH-box helicase 33
There are no transcripts associated with this gene.

Gene
RPAIN
Gene Name
RPA interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033002.3 Intron NP_001028174.2
NM_001160243.1 Intron NP_001153715.1
NM_001160244.1 Intron NP_001153716.1
NM_001160246.1 Intron NP_001153718.1
NM_001160266.1 Intron NP_001153738.1

View Full Product Details