Product Details

SNP ID
hCV86381537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29256100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTAGCAAAAGGGAAAATTTCT[G/T]GTGACTGTTCTTATATGTAGAAAAG
Phenotype
MIM: 123610 MIM: 609356
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CRYBA1 PubMed Links

Gene Details

Gene
CRYBA1
Gene Name
crystallin beta A1
There are no transcripts associated with this gene.

Gene
NUFIP2
Gene Name
NUFIP2, FMR1 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020772.2 10616 UTR 3 NP_065823.1

View Full Product Details