Product Details

SNP ID
hCV86387813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43253628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCAGTCATGACTGAGACTCTGTC[A/T]GGTCTCCATGGCAGGGACCTGATTG
Phenotype
MIM: 176398
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC284344 PubMed Links

Gene Details

Gene
LOC284344
Gene Name
uncharacterized LOC284344
There are no transcripts associated with this gene.

Gene
PSG9
Gene Name
pregnancy specific beta-1-glycoprotein 9
There are no transcripts associated with this gene.

View Full Product Details