Product Details

SNP ID
hCV86388493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50517827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCCGCGGCGGCCACGGACACG[C/G]CCGAGGCGGCGGGCAGCTCGTCCTC
Phenotype
MIM: 615324
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASPDH PubMed Links

Gene Details

Gene
ASPDH
Gene Name
aspartate dehydrogenase domain containing
There are no transcripts associated with this gene.

Gene
JOSD2
Gene Name
Josephin domain containing 2
There are no transcripts associated with this gene.

Gene
LRRC4B
Gene Name
leucine rich repeat containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080457.1 2023 Missense Mutation GCC,GGC A629G NP_001073926.1

View Full Product Details