Product Details

SNP ID
rs41345851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23083142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGTAGTCAAGTGTGTGGTAACCT[C/T]AGTACATTTGGATGAATTAATTCAA
Phenotype
MIM: 120577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD93 PubMed Links

Gene Details

Gene
CD93
Gene Name
CD93 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012072.3 2915 UTR 3 NP_036204.2

View Full Product Details