Product Details

SNP ID
rs41279795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:45344520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTGTGAGTATTAGAGTGGGAA[C/G]TGAACAGTGATCAGGTGACTGCTGC
Phenotype
MIM: 608685
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM118A PubMed Links

Gene Details

Gene
FAM118A
Gene Name
family with sequence similarity 118 member A
There are no transcripts associated with this gene.

Gene
SMC1B
Gene Name
structural maintenance of chromosomes 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291501.1 3796 UTR 3 NP_001278430.1
NM_148674.4 3796 UTR 3 NP_683515.4
XM_011530144.2 3796 UTR 3 XP_011528446.1
XM_011530145.2 3796 Intron XP_011528447.1

View Full Product Details