Product Details

SNP ID
rs41307371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73822218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCATTTTTCATGAGGGATGGAC[G/T]AGGAAAATGAAGTGTCTATTATAAG
Phenotype
MIM: 300181 MIM: 314670
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TSIX PubMed Links

Gene Details

Gene
TSIX
Gene Name
TSIX transcript, XIST antisense RNA
There are no transcripts associated with this gene.

Gene
XIST
Gene Name
X inactive specific transcript (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details