Product Details

SNP ID
rs41314149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:151408567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCTGTCAAGCACTCTGTGTCA[A/C]CCACCAGATTAGTATAACTATTAAT
Phenotype
MIM: 300913
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
VMA21 PubMed Links

Gene Details

Gene
VMA21
Gene Name
VMA21 vacuolar H+-ATPase homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017980.3 4265 UTR 3 NP_001017980.1
XM_011531125.2 4265 UTR 3 XP_011529427.1

View Full Product Details