Product Details
- SNP ID
-
rs41305207
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:20008468 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAGTATTGTGCAGTTTGTAGCAC[C/T]ATTCTCTCACTAAAACGATCACAAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAP7D2
PubMed Links
Gene Details
- Gene
- MAP7D2
- Gene Name
- MAP7 domain containing 2
- Gene
- MIR23C
- Gene Name
- microRNA 23c
There are no transcripts associated with this gene.
View Full Product Details