Product Details

SNP ID
rs41297313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23000279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGGATCAAAAATAAAAGATCT[A/C]CAACATTCGACAAACACTAAAATAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DDX53 PubMed Links

Gene Details

Gene
DDX53
Gene Name
DEAD-box helicase 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182699.3 319 Silent Mutation CTA,CTC L74L NP_874358.2
Gene
PTCHD1-AS
Gene Name
PTCHD1 antisense RNA (head to head)
There are no transcripts associated with this gene.

View Full Product Details