Product Details

SNP ID
rs41402346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30308289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAAAGAAAGCACTTGATGGCTT[A/G]GACCTGGGAGGCGGAGGGCACCTTC
Phenotype
MIM: 300473
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NR0B1 PubMed Links

Gene Details

Gene
NR0B1
Gene Name
nuclear receptor subfamily 0 group B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000475.4 1090 Nonsense Mutation CAA,TAA Q359* NP_000466.2
XM_017029338.1 1090 Nonsense Mutation CAA,TAA Q359* XP_016884827.1

View Full Product Details