Product Details

SNP ID
rs41300279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:132628946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCCAGACCAGTCATCGCCAGT[A/G]TAGCAGCTGGGCAGCTCTTCGGAGG
Phenotype
MIM: 300545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HS6ST2 PubMed Links

Gene Details

Gene
HS6ST2
Gene Name
heparan sulfate 6-O-sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077188.1 1631 Silent Mutation TAC,TAT Y405Y NP_001070656.1
NM_147175.3 1631 Silent Mutation TAC,TAT Y365Y NP_671704.3
XM_005262490.3 1631 Silent Mutation TAC,TAT Y405Y XP_005262547.1
XM_005262491.2 1631 Silent Mutation TAC,TAT Y376Y XP_005262548.1
XM_011531406.1 1631 Silent Mutation TAC,TAT Y259Y XP_011529708.1
XM_011531407.2 1631 Intron XP_011529709.1
XM_011531408.2 1631 Intron XP_011529710.1
XM_017029944.1 1631 Silent Mutation TAC,TAT Y365Y XP_016885433.1
XM_017029945.1 1631 Silent Mutation TAC,TAT Y259Y XP_016885434.1
XM_017029946.1 1631 Silent Mutation TAC,TAT Y230Y XP_016885435.1

View Full Product Details