Product Details

SNP ID
rs41313989
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:117111461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTAATCTTCCAGGCTTTGGGGCA[C/G]AAGGCTGTCATGGCATAGAGACGGA
Phenotype
MIM: 609318 MIM: 604718
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRIM45 PubMed Links

Gene Details

Gene
TRIM45
Gene Name
tripartite motif containing 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145635.1 3175 UTR 3 NP_001139107.1
NM_025188.3 3175 UTR 3 NP_079464.2
XM_011542199.2 3175 UTR 3 XP_011540501.1
Gene
TTF2
Gene Name
transcription termination factor 2
There are no transcripts associated with this gene.

View Full Product Details