Product Details

SNP ID
rs41302821
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:109506542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTCTGTGGCAGGCATCCATAAAA[G/T]CTCTGCCTTCAGCAGCCCCCACTCT
Phenotype
MIM: 615689
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AMIGO1 PubMed Links

Gene Details

Gene
AMIGO1
Gene Name
adhesion molecule with Ig like domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020703.2 2759 Intron NP_065754.2
XM_011541812.2 2759 UTR 3 XP_011540114.1
Gene
CYB561D1
Gene Name
cytochrome b561 family member D1
There are no transcripts associated with this gene.

View Full Product Details