Product Details
- SNP ID
-
rs45455292
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:111414948 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGGTCAGGGTCTTTTCCCCAGGG[C/G]TCACAGACTGATAACCCACAGAGGT
- Phenotype
-
MIM: 603578
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OVGP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs549398942] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OVGP1
- Gene Name
- oviductal glycoprotein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002557.3 |
1609 |
Missense Mutation |
ACC,AGC |
T518S |
NP_002548.3 |
View Full Product Details