Product Details

SNP ID
rs41317063
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:207758165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGTACTGAGCCCTCGGTAATA[A/G]CCTTTGTTCTGTAGGGTTTTGAAGT
Phenotype
MIM: 120920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD46 PubMed Links

Gene Details

Gene
CD46
Gene Name
CD46 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002389.4 Intron NP_002380.3
NM_153826.3 Intron NP_722548.1
NM_172350.2 Intron NP_758860.1
NM_172351.2 Intron NP_758861.1
NM_172352.2 Intron NP_758862.1
NM_172353.2 Intron NP_758863.1
NM_172359.2 Intron NP_758869.1
NM_172361.2 Intron NP_758871.1
XM_011509563.1 Intron XP_011507865.1
XM_011509564.1 Intron XP_011507866.1
XM_017001308.1 Intron XP_016856797.1
XM_017001309.1 Intron XP_016856798.1
XM_017001310.1 Intron XP_016856799.1

View Full Product Details