Product Details

SNP ID
rs45625937
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:6460882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCAACTGCTGCACCCCATCCC[G/T]ACCCTGTCTCCGCCACCTCGCAGCC
Phenotype
MIM: 606351 MIM: 611101 MIM: 603366
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ESPN PubMed Links

Gene Details

Gene
ESPN
Gene Name
espin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031475.2 3469 UTR 3 NP_113663.2
XM_005263501.2 3469 UTR 3 XP_005263558.1
XM_011542231.1 3469 Intron XP_011540533.1
XM_011542232.1 3469 Intron XP_011540534.1
XM_011542233.2 3469 Intron XP_011540535.1
XM_011542236.2 3469 Intron XP_011540538.1
XM_011542237.1 3469 Intron XP_011540539.1
XM_011542238.2 3469 Intron XP_011540540.1
XM_017002433.1 3469 Intron XP_016857922.1
XM_017002434.1 3469 Intron XP_016857923.1
Gene
PLEKHG5
Gene Name
pleckstrin homology and RhoGEF domain containing G5
There are no transcripts associated with this gene.

Gene
TNFRSF25
Gene Name
TNF receptor superfamily member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039664.1 3469 Intron NP_001034753.1
NM_003790.2 3469 Intron NP_003781.1
NM_148965.1 3469 Intron NP_683866.1
NM_148966.1 3469 Intron NP_683867.1
NM_148967.1 3469 Intron NP_683868.1
NM_148970.1 3469 Intron NP_683871.1

View Full Product Details