Product Details

SNP ID
rs45514194
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:8012028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTATCTCTTCAAGATAAATACA[C/T]GGAAGGATGTGAAAATCGGAACACC
Phenotype
MIM: 608069
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERRFI1 PubMed Links

Gene Details

Gene
ERRFI1
Gene Name
ERBB receptor feedback inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018948.3 2834 UTR 3 NP_061821.1
XM_005263477.2 2834 UTR 3 XP_005263534.1
XM_006710697.2 2834 UTR 3 XP_006710760.1
XM_011541596.2 2834 UTR 3 XP_011539898.1

View Full Product Details