Product Details

SNP ID
rs41274498
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:11066640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTTCTTCAGGAAGAATTTTAAT[C/G]GTTTAAAAATATGTATGTACAAAAG
Phenotype
MIM: 605960 MIM: 182891
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EXOSC10 PubMed Links

Gene Details

Gene
EXOSC10
Gene Name
exosome component 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001998.2 2815 UTR 3 NP_001001998.1
NM_002685.3 2815 UTR 3 NP_002676.1
XM_005263475.2 2815 UTR 3 XP_005263532.2
XM_017001494.1 2815 UTR 3 XP_016856983.1
XM_017001495.1 2815 UTR 3 XP_016856984.1
XM_017001496.1 2815 UTR 3 XP_016856985.1
XM_017001497.1 2815 UTR 3 XP_016856986.1
Gene
SRM
Gene Name
spermidine synthase
There are no transcripts associated with this gene.

View Full Product Details