Product Details

SNP ID
rs45572531
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:11790692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGACGCAGGGTCATGGAGCCTC[T/C]GTTTCTCTCGCATTCTGGGTGGGCC
Phenotype
MIM: 607093
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
C1orf167 PubMed Links

Gene Details

Gene
C1orf167
Gene Name
chromosome 1 open reading frame 167
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010881.1 3025 Intron NP_001010881.1
Gene
MTHFR
Gene Name
methylenetetrahydrofolate reductase (NAD(P)H)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005957.4 3025 Silent Mutation ACA,ACG T653T NP_005948.3
XM_005263458.3 3025 Silent Mutation ACA,ACG T694T XP_005263515.1
XM_005263460.4 3025 Silent Mutation ACA,ACG T653T XP_005263517.1
XM_005263462.4 3025 Silent Mutation ACA,ACG T653T XP_005263519.1
XM_005263463.3 3025 Silent Mutation ACA,ACG T571T XP_005263520.1
XM_011541495.2 3025 Silent Mutation ACA,ACG T693T XP_011539797.1
XM_011541496.2 3025 Missense Mutation AGA,GGA R634G XP_011539798.1
XM_017001328.1 3025 Silent Mutation ACA,ACG T644T XP_016856817.1

View Full Product Details