Product Details

SNP ID
rs41294736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112774370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGCTATTGTGGAGGATGCCTG[C/G]TCACACTCAGAGGATGAAGGGGTTC
Phenotype
MIM: 616174 MIM: 147760
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CKAP2L PubMed Links

Gene Details

Gene
CKAP2L
Gene Name
cytoskeleton associated protein 2 like
There are no transcripts associated with this gene.

Gene
IL1A
Gene Name
interleukin 1 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000575.4 2477 UTR 3 NP_000566.3

View Full Product Details