Product Details

SNP ID
rs45542334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136440363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGATCCAGCGGGACACGCCTGTG[A/G]AACATTTTGGAAAACATCCTGGGTG
Phenotype
MIM: 613037 MIM: 612854
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INPP5E PubMed Links

Gene Details

Gene
INPP5E
Gene Name
inositol polyphosphate-5-phosphatase E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318502.1 8620 Intron NP_001305431.1
NM_019892.5 8620 Intron NP_063945.2
XM_017014926.1 8620 Intron XP_016870415.1
Gene
SEC16A
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276418.1 8620 UTR 3 NP_001263347.1
NM_014866.1 8620 UTR 3 NP_055681.1
XM_005266134.2 8620 UTR 3 XP_005266191.1
XM_011519246.2 8620 UTR 3 XP_011517548.1
XM_011519247.1 8620 UTR 3 XP_011517549.1
XM_011519248.2 8620 UTR 3 XP_011517550.1
XM_011519249.1 8620 UTR 3 XP_011517551.1
XM_011519250.2 8620 Intron XP_011517552.1
XM_011519251.1 8620 UTR 3 XP_011517553.1
XM_011519252.2 8620 UTR 3 XP_011517554.1
XM_011519253.1 8620 UTR 3 XP_011517555.1
XM_011519254.1 8620 UTR 3 XP_011517556.1
XM_011519255.2 8620 UTR 3 XP_011517557.1
XM_011519256.1 8620 UTR 3 XP_011517558.1
XM_011519257.1 8620 UTR 3 XP_011517559.1
XM_011519258.1 8620 UTR 3 XP_011517560.1
XM_011519259.1 8620 UTR 3 XP_011517561.1
XM_011519260.2 8620 UTR 3 XP_011517562.1
XM_011519261.1 8620 UTR 3 XP_011517563.1
XM_011519262.1 8620 UTR 3 XP_011517564.1
XM_011519263.2 8620 UTR 3 XP_011517565.1
XM_011519264.2 8620 Intron XP_011517566.1
XM_017015363.1 8620 UTR 3 XP_016870852.1
XM_017015364.1 8620 Intron XP_016870853.1

View Full Product Details