Product Details

SNP ID
rs41289997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137879312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGGTAGGCCTGGCAGATAACA[C/G]CTTCCTCGAGGGCCAGGGCAGCTGG
Phenotype
MIM: 601012
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CACNA1B PubMed Links

Gene Details

Gene
CACNA1B
Gene Name
calcium voltage-gated channel subunit alpha1 B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000718.3 Intron NP_000709.1
NM_001243812.1 Intron NP_001230741.1
XM_011518991.2 Intron XP_011517293.1
XM_011518992.2 Intron XP_011517294.1
XM_011518993.2 Intron XP_011517295.1
XM_011518994.2 Intron XP_011517296.1
XM_017015115.1 Intron XP_016870604.1
XM_017015116.1 Intron XP_016870605.1
XM_017015117.1 Intron XP_016870606.1
XM_017015118.1 Intron XP_016870607.1
XM_017015119.1 Intron XP_016870608.1
XM_017015120.1 Intron XP_016870609.1
Gene
LOC100133077
Gene Name
uncharacterized LOC100133077
There are no transcripts associated with this gene.

View Full Product Details