Product Details

SNP ID
rs41298391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132570527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAGCTGCACTCTCCGACCAATT[C/T]TCTCGTTGCCTCTCTGGCCTGCGCT
Phenotype
MIM: 608923
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAAR6 PubMed Links

Gene Details

Gene
TAAR6
Gene Name
trace amine associated receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175067.1 206 Missense Mutation TCT,TTT S69F NP_778237.1

View Full Product Details