Product Details

SNP ID
rs45492697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:30628188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTAAAAAACAATGACAATATTT[G/T]ATAAAACTTTTTATTTTCAGTGGCT
Phenotype
MIM: 611299
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
G2E3 PubMed Links

Gene Details

Gene
G2E3
Gene Name
G2/M-phase specific E3 ubiquitin protein ligase
There are no transcripts associated with this gene.

Gene
SCFD1
Gene Name
sec1 family domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257376.1 Intron NP_001244305.1
NM_001283031.1 Intron NP_001269960.1
NM_001283032.1 Intron NP_001269961.1
NM_001283033.1 Intron NP_001269962.1
NM_016106.3 Intron NP_057190.2
NM_182835.2 Intron NP_878255.1
XM_005267469.2 Intron XP_005267526.1
XM_011536586.2 Intron XP_011534888.1
XM_011536587.2 Intron XP_011534889.1
XM_011536588.2 Intron XP_011534890.1
XM_011536589.1 Intron XP_011534891.1
XM_017021113.1 Intron XP_016876602.1

View Full Product Details