Product Details

SNP ID
rs41463347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77343218 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCTTCCTCTTCTTCATCCTCATC[A/G]TCACTGGAATCACTGACCTGCACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMED8 PubMed Links

Gene Details

Gene
TMED8
Gene Name
transmembrane p24 trafficking protein family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213601.1 619 Silent Mutation GAC,GAT D240D NP_998766.1
XM_017021223.1 619 Silent Mutation GAC,GAT D347D XP_016876712.1
XM_017021224.1 619 Silent Mutation GAC,GAT D177D XP_016876713.1
XM_017021225.1 619 Silent Mutation GAC,GAT D172D XP_016876714.1

View Full Product Details