Product Details

SNP ID
rs45572135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:87934023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGAAAAACGTTCACAGAGAGT[G/T]ATAGTGGTTACAAGACCAAAACTTG
Phenotype
MIM: 606890
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GALC PubMed Links

Gene Details

Gene
GALC
Gene Name
galactosylceramidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000153.3 3156 UTR 3 NP_000144.2
NM_001201401.1 3156 UTR 3 NP_001188330.1
NM_001201402.1 3156 UTR 3 NP_001188331.1
XM_011536618.2 3156 UTR 3 XP_011534920.1

View Full Product Details