Product Details

SNP ID
rs45438692
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35617211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTGGATCTGAGGCAGGAGATGC[C/T]TCCCCCGCGGGTGTTCAAGAGCTTT
Phenotype
MIM: 604205 MIM: 603038
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CPNE1 PubMed Links

Gene Details

Gene
CPNE1
Gene Name
copine 1
There are no transcripts associated with this gene.

Gene
FER1L4
Gene Name
fer-1 like family member 4, pseudogene
There are no transcripts associated with this gene.

Gene
SPAG4
Gene Name
sperm associated antigen 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317931.1 146 Missense Mutation CCT,CTT P50L NP_001304860.1
NM_003116.2 146 Missense Mutation CCT,CTT P127L NP_003107.1
XM_005260520.4 146 Missense Mutation CCT,CTT P34L XP_005260577.1
XM_011529009.2 146 Missense Mutation CCT,CTT P83L XP_011527311.1

View Full Product Details