Product Details

SNP ID
rs41299368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196050240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTATAAATGTATGCTACTATCTC[A/G]TTAAGTAGAGGACCTGGAGAAACCA
Phenotype
MIM: 190010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TFRC PubMed Links

Gene Details

Gene
TFRC
Gene Name
transferrin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128148.2 3729 UTR 3 NP_001121620.1
NM_001313965.1 3729 UTR 3 NP_001300894.1
NM_001313966.1 3729 UTR 3 NP_001300895.1
NM_003234.3 3729 UTR 3 NP_003225.2

View Full Product Details