Product Details

SNP ID
rs45559436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:47300780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGGCCCCCGCCTGGTCCGCACT[G/A]CCCGCGGCCGCCGACGGCCTGCAGG
Phenotype
MIM: 601361
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
GDF10 PubMed Links

Gene Details

Gene
GDF10
Gene Name
growth differentiation factor 10
There are no transcripts associated with this gene.

View Full Product Details