Product Details

SNP ID
rs41301052
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:42430770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATGGTCTTCAGTTGAAATGAGA[A/C]TTAATAACTCTTGAATGGTGCTTTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RIMKLA PubMed Links

Gene Details

Gene
RIMKLA
Gene Name
ribosomal modification protein rimK like family member A
There are no transcripts associated with this gene.

Gene
ZMYND12
Gene Name
zinc finger MYND-type containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146192.1 1334 Missense Mutation AGT,ATT S245I NP_001139664.1
NM_032257.4 1334 Missense Mutation AGT,ATT S355I NP_115633.3

View Full Product Details