Product Details

SNP ID
rs41293279
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:46340907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACGGTCCCGCGGAGACATCGATA[C/T]AAGAAGAAATGGGTAAGGTCCGGCT
Phenotype
MIM: 615394
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NSUN4 PubMed Links

Gene Details

Gene
NSUN4
Gene Name
NOP2/Sun RNA methyltransferase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256127.1 731 Intron NP_001243056.1
NM_001256128.1 731 Intron NP_001243057.1
NM_199044.3 731 Silent Mutation TAC,TAT Y27Y NP_950245.2

View Full Product Details