Product Details

SNP ID
rs41264003
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:32247432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTCAAACCTGCCATGTCCCTGG[A/G]GCTACTGAAATTCCAGGCAGTGGGT
Phenotype
MIM: 153390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM167B PubMed Links

Gene Details

Gene
FAM167B
Gene Name
family with sequence similarity 167 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032648.2 216 Missense Mutation GAG,GGG E4G NP_116037.2
Gene
LCK
Gene Name
LCK proto-oncogene, Src family tyrosine kinase
There are no transcripts associated with this gene.

Gene
MTMR9LP
Gene Name
myotubularin related protein 9-like, pseudogene
There are no transcripts associated with this gene.

View Full Product Details