Product Details

SNP ID
rs41270783
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:38874832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTAGAGGAACCCCATGTAGCTC[G/T]AAGCCACCGCGGTTTCCAATCGCAG
Phenotype
MIM: 611923 MIM: 606535
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GJA9 PubMed Links

Gene Details

Gene
GJA9
Gene Name
gap junction protein alpha 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030772.4 1557 Silent Mutation NP_110399.2
Gene
GJA9-MYCBP
Gene Name
GJA9-MYCBP readthrough
There are no transcripts associated with this gene.

Gene
LOC105378663
Gene Name
uncharacterized LOC105378663
There are no transcripts associated with this gene.

Gene
MYCBP
Gene Name
MYC binding protein
There are no transcripts associated with this gene.

View Full Product Details