Product Details

SNP ID
rs41268213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:70203552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGTTCCTATGCCTCGGGCGCGT[G/T]GGCAAGCGCAAGCTGGAGCGCGTGC
Phenotype
MIM: 602457
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FADD PubMed Links

Gene Details

Gene
FADD
Gene Name
Fas associated via death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003824.3 390 Silent Mutation GTG,GTT V31V NP_003815.1

View Full Product Details