Product Details

SNP ID
rs41300562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125709697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAATGTAACATTTTAATTTATT[A/G]ACTGATAATTAGTTCTATTTTCATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDRF1 PubMed Links

Gene Details

Gene
EDRF1
Gene Name
erythroid differentiation regulatory factor 1
There are no transcripts associated with this gene.

Gene
FLJ37035
Gene Name
uncharacterized LOC399821
There are no transcripts associated with this gene.

Gene
LOC283038
Gene Name
uncharacterized LOC283038
There are no transcripts associated with this gene.

View Full Product Details