Product Details

SNP ID
rs45592334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:532310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGGCCCACAGAGGCCTGGGAGG[A/G]GAGCTAAGGGCTGGGGTTCCGGTGG
Phenotype
MIM: 190020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HRAS PubMed Links

Gene Details

Gene
HRAS
Gene Name
HRas proto-oncogene, GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130442.2 993 UTR 3 NP_001123914.1
NM_001318054.1 993 UTR 3 NP_001304983.1
NM_005343.3 993 UTR 3 NP_005334.1
NM_176795.4 993 UTR 3 NP_789765.1
Gene
LOC101059906
Gene Name
collagen alpha-2(IV) chain-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_003959933.4 993 Intron XP_003959982.2
Gene
LRRC56
Gene Name
leucine rich repeat containing 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198075.3 993 Intron NP_932341.1
XM_005252775.2 993 Intron XP_005252832.1
XM_005252776.2 993 Intron XP_005252833.1
XM_006718132.2 993 Intron XP_006718195.1
XM_006718133.2 993 Intron XP_006718196.1
XM_011519875.2 993 Intron XP_011518177.1
XM_011519876.1 993 Intron XP_011518178.1
XM_011519877.2 993 Intron XP_011518179.1
XM_017017167.1 993 Intron XP_016872656.1
XM_017017168.1 993 Intron XP_016872657.1

View Full Product Details