Product Details

SNP ID
rs57350253
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:151134568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCCTGGGATGGTGTCTGTGGCC[A/G]TCATGGCCATGTGCAGCTTTGGCTG
Phenotype
MIM: 609294
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABPB2 PubMed Links

Gene Details

Gene
GABPB2
Gene Name
GA binding protein transcription factor beta subunit 2
There are no transcripts associated with this gene.

Gene
SEMA6C
Gene Name
semaphorin 6C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178061.1 Intron NP_001171532.1
NM_001178062.1 Intron NP_001171533.1
NM_030913.4 Intron NP_112175.2
XM_005244835.3 Intron XP_005244892.1
XM_017000075.1 Intron XP_016855564.1
XM_017000076.1 Intron XP_016855565.1
XM_017000077.1 Intron XP_016855566.1
XM_017000078.1 Intron XP_016855567.1
XM_017000079.1 Intron XP_016855568.1
XM_017000080.1 Intron XP_016855569.1
XM_017000081.1 Intron XP_016855570.1
XM_017000082.1 Intron XP_016855571.1

View Full Product Details