Product Details

SNP ID
rs61814149
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:170936119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAATCTCCTAACTTCATGCCTTA[C/T]TAGTAAGGATTATCTTATTAGTAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MROH9 PubMed Links

Gene Details

Gene
MROH9
Gene Name
maestro heat like repeat family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163629.1 Intron NP_001157101.1
NM_025063.2 Intron NP_079339.2
XM_011510005.2 Intron XP_011508307.1
XM_011510006.2 Intron XP_011508308.1
XM_011510007.2 Intron XP_011508309.1

View Full Product Details