Product Details

SNP ID
rs61747503
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:172442315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCCGCCCTTCACCTCCATCA[A/G]TGAGATCAAACAAAACATACCCAAT
Phenotype
MIM: 601730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf105 PubMed Links

Gene Details

Gene
C1orf105
Gene Name
chromosome 1 open reading frame 105
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300760.1 Intron NP_001287689.1
NM_139240.3 Intron NP_640333.3
XM_005245608.2 Intron XP_005245665.1
XM_005245610.3 Intron XP_005245667.1
XM_006711630.2 Intron XP_006711693.1
XM_011510152.2 Intron XP_011508454.1
XM_011510153.2 Intron XP_011508455.1
XM_011510154.2 Intron XP_011508456.1
XM_011510155.2 Intron XP_011508457.1
XM_011510156.2 Intron XP_011508458.1
XM_017002808.1 Intron XP_016858297.1
Gene
PIGC
Gene Name
phosphatidylinositol glycan anchor biosynthesis class C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002642.3 Intron NP_002633.1
NM_153747.1 Intron NP_714969.1

View Full Product Details