Product Details

SNP ID
rs58733562
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:152698960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCTCCCAAGTGCCCCCCAAAAT[A/G]CCCACCCAAGTGTCCTCCAAAGTGC
Phenotype
MIM: 612609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCE2A PubMed Links

Gene Details

Gene
LCE2A
Gene Name
late cornified envelope 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178428.3 110 Missense Mutation TAC,TGC Y20C NP_848515.1

View Full Product Details