Product Details

SNP ID
rs61807501
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:182850014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTTTTTTTTTAATTTCATCAG[C/T]GTTCAGCTTTATTGATAACATCCTC
Phenotype
MIM: 603115
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHX9 PubMed Links

Gene Details

Gene
DHX9
Gene Name
DEAH-box helicase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001357.4 Intron NP_001348.2

View Full Product Details