Product Details

SNP ID
rs58166377
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:202012104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATGGATGGCGCCACCCTCTGCA[A/G]TTGTGCCCTTGAGGAGCTGCGTCTG
Phenotype
MIM: 602191 MIM: 602675
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELF3 PubMed Links

Gene Details

Gene
ELF3
Gene Name
E74 like ETS transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114309.1 464 Missense Mutation AAT,AGT N104S NP_001107781.1
NM_004433.4 464 Missense Mutation AAT,AGT N104S NP_004424.3
XM_005244942.2 464 Missense Mutation AAT,AGT N104S XP_005244999.1
Gene
MIR6740
Gene Name
microRNA 6740
There are no transcripts associated with this gene.

Gene
RNPEP
Gene Name
arginyl aminopeptidase
There are no transcripts associated with this gene.

View Full Product Details