Product Details

SNP ID
hCV88356475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:203083181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAAAAGGAAATCCAAATAAGTT[A/T]GCAAAAAAAAAAAAAAAAAAAAAAA
Phenotype
MIM: 159980 MIM: 603145
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MYOG PubMed Links

Gene Details

Gene
MYOG
Gene Name
myogenin (myogenic factor 4)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002479.5 1481 UTR 3 NP_002470.2
Gene
PPFIA4
Gene Name
PTPRF interacting protein alpha 4
There are no transcripts associated with this gene.

View Full Product Details